Merck
AXYSCT050RS
Screw cap tubes and caps with O-rings, conical bottom
capacity 0.5 mL, red, sterile
Synonym(e):
centrifuge tubes
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.22
Materialien
red
screw top cap
Sterilität
sterile
Leistungsmerkmale
RCF 20,000 × g
Verpackung
pack of 500 tubes
case of 8 packs
Hersteller/Markenname
Axygen Inc
Corning
Kapazität
0.5 mL