AXYRESV25S
Reservoir
capacity 25 mL, sterile
Synonym(e):
Liquid Reservoir, Reservoir Container, Storage Reservoir
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121522
NACRES:
NB.13
Empfohlene Produkte
Sterilität
sterile
Hersteller/Markenname
Axygen Inc
Corning
Kapazität
25 mL
Verwandte Kategorien
Allgemeine Beschreibung
- Axygen® reagent reservoirs are optimized for use with multichannel pipettors.
- The pour-off spouts on each corner and the sharp "V" bottom design facilitate maximum sample retrieval.
Rechtliche Hinweise
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)