Merck
AXYP96450VCS
V-bottom assay plate
size 96 wells, V-bottom clear polypropylene wells, maximum volume 500 μL, pkg of 10x5plates/cs, sterile
Synonym(e):
V-Bottom Plate
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.22
Materialien:
V-bottom clear polypropylene wells
clear plate
Maximales Volumen:
500 μL
Größe:
96 wells
Arbeitsvolumen:
450 μL
Materialien
V-bottom clear polypropylene wells
clear plate
Sterilität
sterile
Leistungsmerkmale
lid: no
skirt
Hersteller/Markenname
Axygen Inc P-96-450V-C-S
Corning
Verpackung
pkg of 10x5plates/cs
Maximales Volumen
500 μL
Größe
96 wells
Arbeitsvolumen
450 μL
Bindungsart
non-treated surface
Allgemeine Beschreibung
  • Axygen® V-bottom assay plates are manufactured using chemically resistant polypropylene.
  • The plates can withstand temperatures down to -80°C.
  • Ideal for sample collection, in vitro growth chambers, and long-term storage.
  • The ultra-flat surface of the plates can be sealed using heat sealing film for sample protection.
Rechtliche Hinweise
Axygen is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)