Merck
ADF356302S-EP
Adapter for Eppendorf® F-35-6-30 Rotor
holds 1 x 2.6 - 7 mL round-bottom tube, small rotor bore, small rotor bore, pack of 2 ea
About This Item
UNSPSC-Code:
41103911
Leistungsmerkmale
holds 1 x 2.6 - 7 mL round-bottom tube, small rotor bore
Verpackung
pack of 2 ea
Hersteller/Markenname
Eppendorf® 022654501 (NA)
Methode(n)
centrifugation: suitable
Kompatibilität
for use with Rotor F-35-6-30 (small rotor bore)
Allgemeine Beschreibung
Adapters to fit various size tubes and bottles for use with Eppendorf rotor F-35-6-30.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)