Merck
01-1730
Sigma-Aldrich
Adipoylchlorid
SAJ first grade, ≥98.0 %
Synonym(e):
Adipylchlorid
About This Item
Lineare Formel:
ClCO(CH2)4COCl
CAS-Nummer:
Molekulargewicht:
183.03
Beilstein:
507709
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
12352101
PubChem Substanz-ID:
Qualität
SAJ first grade
Verfügbarkeit
available only in Japan
Brechungsindex
n20/D 1.471 (lit.)
Kapazität
≥98.0 %
bp
105-107 °C/2 mmHg (lit.)
Dichte
1.259 g/mL at 25 °C (lit.)
SMILES String
ClC(=O)CCCCC(Cl)=O
InChI
1S/C6H8Cl2O2/c7-5(9)3-1-2-4-6(8)10/h1-4H2
InChIKey
PWAXUOGZOSVGBO-UHFFFAOYSA-N
Piktogramme
Corrosion
Signalwort
Danger
H-Sätze
Gefahreneinstufungen
Skin Corr. 1B
Lagerklassenschlüssel
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flammpunkt (°F)
233.6 °F - closed cup
Flammpunkt (°C)
112 °C - closed cup
Persönliche Schutzausrüstung
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Nicolò Maccaferri et al.
Nature communications, 6, 6150-6150 (2015-02-03)
Systems allowing label-free molecular detection are expected to have enormous impact on biochemical sciences. Research focuses on materials and technologies based on exploiting localized surface plasmon resonances in metallic nanostructures. The reason for this focused attention is their suitability for
Jungeun Bae et al.
The AAPS journal, 17(6), 1357-1368 (2015-07-01)
A novel redox-responsive amphiphilic polymer was synthesized with bioreductive trimethyl-locked quinone propionic acid for a potential triggered drug delivery application. The aim of this study was to synthesize and characterize the redox-responsive amphiphilic block copolymer micelles containing pendant bioreductive quinone
Michelle A Ouimet et al.
Macromolecular bioscience, 15(3), 342-350 (2014-10-22)
Polymers such as poly(N-vinyl-2-pyrrolidone) (PVP) have been used to prepare hydrogels for wound dressing applications but are not inherently bioactive. For enhanced healing, PVP was blended with salicylic acid-based poly(anhydride-esters) (SAPAE) and shown to exhibit hydrogel properties upon swelling. In