Wichtige Dokumente
SAU0200L41DP5
Mobius®, Silber, 200 L Standard PureFlex™ Trommel
200L Standard PureFlex™ Drum Assembly
Synonym(e):
Mobius® Silber Trommelanordnung
Alle Fotos(1)
About This Item
UNSPSC-Code:
23151806
eCl@ss:
32031690
Empfohlene Produkte
Materialien
PureFlex™ container
Qualitätsniveau
Produktlinie
EMPROVE® Single Use
Mobius®
Parameter
200 L sample volume
Rechtliche Hinweise
Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany
PUREFLEX is a trademark of Merck KGaA, Darmstadt, Germany
Global Trade Item Number
SKU | GTIN |
---|---|
SAU0200L41DP5 |