Merck
Z758833
Bollé Cobra wrap-around safety spectacles
CE compliant
Synonym(e):
protective eyewear, protective glasses, safety eyeglasses, safety eyewear, safety glasses, safety spectacles
About This Item
UNSPSC-Code:
46181802
NACRES:
NB.35
Leistungsmerkmale
CE compliant
Verpackung
pkg of 1 ea
Hersteller/Markenname
Bollé COBPSI
Verfügbarkeit
not available in USA
Allgemeine Beschreibung
A multi-purpose wrap around safety goggle with a panoramic visual field. Enveloping and without any visual interference, the Cobra offers 180° panoramic visual field with perfect optical quality. The ability to interchange the temples with an adjustable strap and foam option makes the Cobra an ultra versatile model.
Cobra Kits to facilitate the optional strap conversion is available separately as: Z758868 or Z758876.
  • Upper protection
  • Interchangeable comfort foam
  • Anti-scratch, anti-fog, anti-static coatings
  • Non-slip bridge
  • Adjustable strap
Tested and approved to EN 166/170/172 1B CE
Ähnliches Produkt
Produkt-Nr.
Beschreibung
Preisangaben
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z758833-1EA4061833345337