Merck
Z748234
Sigma-Aldrich
Load flask for Aldrich® Kugelrohr apparatus
capacity 250 mL, joint: ST/NS 14/20
Synonym(e):
distillation apparatus, distilling apparatus, fractional distillation apparatus, glass distillation apparatus, kugelrohr apparatus, kugelrohr distillation, short path distillation apparatus, vacuum distillation apparatus
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
glass
single-neck
spherical
Verbindung
joint: ST/NS14/20
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard Taper Joint
Verpackung
case of 1
Hersteller/Markenname
Sigma-Aldrich
Kapazität
250 mL
Allgemeine Beschreibung
Heavy wall glass flask with solid support that attaches to Kugelrohr motor drive coupling. Fill load flask one-third full with distillable material.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)