Merck
Z743191
Julabo® bath test tube racks
to fit 21 tubes, diam. 30 mm, plastic
Synonym(e):
tube holder
About This Item
UNSPSC-Code:
41103706
NACRES:
NB.77
Materialien
plastic
Verpackung
pack of 1 ea
Hersteller/Markenname
Julabo 9970303
Parameter
100 °C max. temp.
Durchmesser
30 mm
Allgemeine Beschreibung
Plastic rack suitable for temperatures up to 100°C. If solvent based samples or acidic samples are tempered above 60°C, it is recommended using test glass inserts made of stainless steel.
Rechtliche Hinweise
Julabo is a registered trademark of Julabo USA, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z743191-1EA