Z743183
Julabo® bath covers
lift-up transparent cover to fit Pura 22
Alle Fotos(1)
About This Item
UNSPSC-Code:
41103706
NACRES:
NB.77
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
Julabo 9970583
Verwandte Kategorien
Allgemeine Beschreibung
Hinged cover for use with PURA 22 water bath, reduces the evaporation of the bath fluid and saves energy. The rising steam is led back into the bath. This means that the bath fluid remains in the bath and refill cycles are extended. An additional benefit is that the cover prevents contamination of the samples in the bath.
Rechtliche Hinweise
Julabo is a registered trademark of Julabo USA, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z743183-1EA |