Merck
Z742867
Kern PLS series precision balances
Kern PLS 420-3F, weighing capacity 420 g, accuracy: 0.001 g, precision: ±0.004 g
Synonym(e):
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
About This Item
UNSPSC-Code:
41111502
NACRES:
NB.71
Aufnahme AC/DC
110 - 240 V AC, universal plug set
Verpackung
pack of 1 ea
Hersteller/Markenname
Kern PLS 420-3F
Messbereich Wiegekapazität
420 g, accuracy: 0.001 g, precision: ±0.004 g
Verwandte Kategorien
Allgemeine Beschreibung
Kern precision balances offer maximum accuracy for demanding measuring tasks in laboratories, industry and research. With versatile functions, simple operation and robust construction, they are the ideal solution for reliable results in any environment. The PLS series of balances offers a high-quality precision scale with comfortable graphic display and enormous weighing range.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)