Z742596
Accuris™ SmartDoc™ narrow band pass filter
590nm filter; Used for imaging EtBR
Synonym(e):
Accuris™ SmartDoc™
Alle Fotos(1)
About This Item
UNSPSC-Code:
41106000
NACRES:
NB.24
Empfohlene Produkte
Leistungsmerkmale
590nm filter; Used for imaging EtBR
Verpackung
pkg of 1
Hersteller/Markenname
Benchmark Scientific E5001-590
Allgemeine Beschreibung
A narrow band pass filter of 590 nm to use for imaging EtBR on a transilluminator
Rechtliche Hinweise
Accuris is a trademark of Benchmark Scientific, Inc.
SmartDoc is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)