Merck
Z742526
Block for digital dry bath and MyBlock
Holds 1 x 96-well PCR plate
About This Item
UNSPSC-Code:
41102404
NACRES:
NB.22
Leistungsmerkmale
Holds 1 x 96-well PCR plate
Verpackung
pack of 1 ea
Hersteller/Markenname
Benchmark Scientific BSWPCR1
Methode(n)
PCR: suitable
Allgemeine Beschreibung
For use with Benchmark Digital Dry Bath
  • Material: high grade aluminum
  • Precision machined to match the tube shape and size, providing unsurpassed temperature uniformity
Rechtliche Hinweise
MyBlock is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)