Merck
Z742269
DURAN® TILT GL 56 Screw Cap
white polypropylene, pack of 10 ea
Synonym(e):
292295602
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Materialien
white polypropylene
Verpackung
pack of 10 ea
Hersteller/Markenname
DURAN® 292295602
Fitting
GL 56 thread
Allgemeine Beschreibung
The ergonomic cap is easy to open and close. Made from non-cytotoxic materials. The wide mouth ensures safe and easy pipetting.
Leistungsmerkmale und Vorteile
• Manufactured from Polypropylene• The ergonomic cap, made from non-cytotoxic materials, is easy to open and close• The wide GL 56 moth ensures safe and easy pipetting and helps avoid contamination
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)