Merck
Z742266
DURAN® GL 56 Bottle Tag
purple silicone
Synonym(e):
292435667
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Materialien
purple silicone
Verpackung
pack of 20 ea
Hersteller/Markenname
DURAN® 292435667
Allgemeine Beschreibung
Bottle tags can be used on their own for color identification or to secure the protective light sheild around the bottle. The GL 56 bottle tags are available in four colors (orange, yellow, blue and purple).
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)