Z741984
Büchi® Rotavapor R-100
cold trap, AC/DC input 100 AC, US 3-pin plug
Synonym(e):
Rotavapor
Alle Fotos(1)
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.43
Empfohlene Produkte
Verbindung
joint SJ24/40
Hersteller/Markenname
Büchi 11100C202
Verpackung
1 ea of
Allgemeine Beschreibung
The rotary evaporator R-100 is a high-quality, entry level Rotavapor to meet the essential needs in classical laboratory applications. It is capable of a manually adjustable rotation speed of 20 to 280 rpm and equipped with an action jack lift for manual lifting of the evaporating flask. Included is the selected condenser, 1 L receiving and evaporating flasks, and a vapor duct. All accessories are included for out of the box usage.
Rechtliche Hinweise
Büchi is a registered trademark of Büchi Labortechnik AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z741984-1EA | 4061838299444 |