Merck
Z741338
Orbi-Shaker CO2 XL orbital shaker with flat mat platform
AC/DC input 230 V AC, EuroPlug
Synonym(e):
horizontal shaker, lab shaker, reciprocating shaker, remote control laboratory shaker
About This Item
UNSPSC-Code:
41103811
NACRES:
NB.73
Hersteller/Markenname
Benchmark Scientific BT4011-E
Orbi-Shaker
Allgemeine Beschreibung
The Orbi-Shaker CO2 XL is designed for shaking, mixing and aeration applications in extreme environments, including CO2 incubators. It can withstand temperatures from 0 to 60°C and humidity levels up to 100%. To avoid contamination, unintentional release of gas or deviations from protocol, all settings are adjusted via the included remote control module.
This remote control module can be stored outside of the chamber and connects to the shaker via flat (1 mm thickness) cable, that easily passes through a port or underneath an incubator door without compromising the seal. This allows for user adjustment of run time or mixing speed without opening the chamber door(s).
The Orbi-Shaker CO2 XL is supplied with a 16 x 16.5 inch non-slip rubber mat platform.
Leistungsmerkmale und Vorteile
A useful addition, the convenient MAGic Clamp platforms (Z741340) are also available for use with Erlenmeyer flasks and test tube racks. The unique magnetic attachment method has been widely recognized as the easiest way to instantly change between flask clamps of different sizes.
Rechtliche Hinweise
MAGic Clamp is a trademark of Benchmark Scientific, Inc.
Orbi-Shaker is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z741338-1EA4061834674269