Merck
Z740678
Sigma-Aldrich
Aldrich® Essentials filter flask, with side arm
capacity 100 mL
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
borosilicate glass flask
flat bottom flask
Verbindung
joint stopper4
Verpackung
pkg of 12 ea
Kapazität
100 mL
Allgemeine Beschreibung
Clear borosilicate glass Erlenmeyer style flask with side arm for connecting to vacuum or aspirator line. Used for filtration with Buchner funnels, Gooch crucibles, etc. Beaded rim for rubber gromets or filter adapters or rubber stoppers.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z740678-12EA4061832120959