Z740242
QianCap™ gas purging set for Ace pressure tubes and bottles
QianCap™-25-v2, fits no. 25 internal thread
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Empfohlene Produkte
Leistungsmerkmale und Vorteile
- Two optional gas feed lines are available.
- Maximum pressure: 60 psi
- Provides optional high-pressure adapter (120 psi).
- Septum for the system is disposable and made of blue PTFE face and white silicone backing.
- Septa fits on Q-Tube-12 mL.
- The blue PTFE liner provides unique chemical resistance and silicon backing has the resealing capability.
Sonstige Hinweise
Pressure flasks/bottles not included
Rechtliche Hinweise
QianCap is a trademark of Q Labtech LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
Z740242-1EA | 4061833405475 |