Merck
Z691577
Mini Bin aluminum foil
W 6 in. (153 mm)
Synonym(e):
aluminum foil, tube foil, tube sealing foil
About This Item
UNSPSC-Code:
41122410
NACRES:
NB.35
Materialien
aluminum film
Sterilität
non-sterile
Verpackung
pack of 1 ea
Hersteller/Markenname
Heathrow Scientific HS23536A
B × L × Dicke
15.3 cm × 152.40 m -0.015 mm
Breite
6 in. (153 mm)
Allgemeine Beschreibung
Designed for easy use and minimal waste. Custom-sized pieces of foil can easily be created for covering small surfaces such as beakers, flasks, tubes and petri dishes. Ideal for autoclaving, freezing, incubating, protecting lab ware, lining, sealing and general storage.
Two sizes available. Designed exclusively for the Mini Bin Foil Dispenser.
Rechtliche Hinweise
Mini Bin is a trademark of Heathrow Scientific
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Slide 1 of 3
1 of 3
Global Trade Item Number
SKUGTIN
Z691577-1EA4061832762814