Z687995
Plas-Labs temperature and humidity control box
AC/DC input 240 V AC, UK plug, Plas-Labs 890-THC/EXP/UK
Alle Fotos(1)
About This Item
UNSPSC-Code:
41123304
NACRES:
NB.35
Empfohlene Produkte
Leistungsmerkmale
CE compliant
Hersteller/Markenname
Plas-Labs 890-THC/EXP/UK
Parameter
16-44 °C temp. range
Innenmaße H × B × T
26 in. × 41 in. × 28 in.
Außenmaße H × B × T
38 in. × 55 in. × 35 in.
Allgemeine Beschreibung
An inert gas source is required for humidity control. One piece clear acrylic formed top, white thermoset bottom with easy to clean corners, and one vacuum capable transfer chamber (12 in. L x 11 in. I.D) with two (2) gas purging valves and one vacuum gauge.
The 8 inch color touchscreen features: one-touch on-screen 24 h datalogging, real time clock, audible alarms, four preset conditions, separate on/off control for temperature
and humidity, and password protected administration window for PID adjustment, test I/O, real time clock adjustment, and alarm conditions.
The 8 inch color touchscreen features: one-touch on-screen 24 h datalogging, real time clock, audible alarms, four preset conditions, separate on/off control for temperature
and humidity, and password protected administration window for PID adjustment, test I/O, real time clock adjustment, and alarm conditions.
- 8 inch color touch screen display
- Thermoelectric(Peltier) assembly
- Nebulizer
- Inert gas hook-up
- One pair of white Hypalon ambidextrous gloves
- Two (2) ground key cock valves for purging
- One hospital grade multiple electric outlet strip
- Fluorescent light and serial port connection fordatalogging.
- Humidity range: 5-95% Rh
Rechtliche Hinweise
Ähnliches Produkt
Produkt-Nr.
Beschreibung
Preisangaben
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)