Z687782
Replacement NMR needle with cone tip for eVol® electronic syringe
for 50 μL, L × O.D. 18.0 cm × 0.63 mm, gauge size 23
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121705
NACRES:
SB.32
Empfohlene Produkte
Manometergröße
23
Hersteller/Markenname
SGE 038038
L × AD
18.0 cm × 0.63 mm
Allgemeine Beschreibung
The eVol® NMR electronic syringe facilitates in-tube sample mixing and enables recovery of precious samples allowing them to be stored in alternative vessels (such as inexpensive vials) rather than being left in valuable tubes. The cone tip replacement needles come in multiple length and gauge options to fit your specific needs.
Rechtliche Hinweise
eVol is a registered trademark of SGE Analytical Science Pty Ltd.
Zubehör
Produkt-Nr.
Beschreibung
Preisangaben
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)