Merck
Z685453
Vacuubrand chemistry pumping units, manual control, Model PC 201 NT
AC/DC input 115 V AC, US 2-pin plug
Synonym(e):
Vacuubrand 20736403, diaphragm pump
About This Item
UNSPSC-Code:
41105101
NACRES:
NB.79
Hersteller/Markenname
Vacuubrand 737003
Parameter
105 mbar vacuum
3.4-3.8 m3/h pumping speed
Allgemeine Beschreibung
This chemistry pumping unit has a wide range of applications like evacuation, evaporation and pumping of gases and vapors in chemical, biological and pharmaceutical laboratories. Typical applications are rotary evaporators, vacuum concentrators and vacuum drying ovens with high boiling solvents.
The manual flow control valve regulates the effective pumping speed at the vacuum connection; the vacuum manometer offers an analog vacuum display. The separator at the inlet (AK), made of glass with a protective coating, retains particles and liquid droplets. The exhaust waste vapor condenser enables near-100-percent solvent recovery.
  • Outstanding chemical resistance and superior vapor tolerance
  • Exceptionally high performance even at low vacuum
  • Excellent ultimate vacuum even with gas ballast
  • Manual vacuum control, analog vacuum display
  • Excellent environmental friendliness due to efficient solvent recovery
Supplied with MD 4C, manual flow control valve, analogue vacuum gauge, inlet catchpot and exhaust waste vapor condenser certification (NRTL).
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)