Z685283
Platinum crucible, with reinforced rim
capacity 25 mL
Synonym(e):
Platinum Dish
Alle Fotos(2)
About This Item
UNSPSC-Code:
41122200
NACRES:
NB.43
Empfohlene Produkte
Hersteller/Markenname
Birmingham metals
Kapazität
25 mL
Allgemeine Beschreibung
Platinum labware has excellent resistance to acids such as acetic, hydrochloric, hydrofluoric, hydroiodic, nitric, selenic, and sulfuric, and to oxidation at high temperatures. These platinum crucibles can be cleaned by boiling in a suitable solvent, for the removal of substances which have alloyed with the surface, scour with alumina powder. Having a reinforced rim means that the rim of the crucible is thicker than the walls to make it stronger; this is where the crucible can be gripped if tongs are used during the handling process.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
Z685283-1EA | 4061833571934 |