Merck
Z682357
Ace rotary evaporator condenser complete "V" assembly
plastic-coated glass, fits Büchi® models R-200/R-205 and R-114/R-144
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
plastic-coated glass
Hersteller/Markenname
Büchi equivalent, 40603/32304
Verwandte Kategorien
Allgemeine Beschreibung
The main glass condenser body with inner coil for vertical installation, O-ring, 1 L receiving flask, stainless steel clamp, GL threaded caps and hose barbs.
Rechtliche Hinweise
Büchi is a registered trademark of Büchi Labortechnik AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z682357-1EA4061832352091