Z677159
NevoCut™ cut protection gloves
size M
Synonym(e):
safety glove
About This Item
Empfohlene Produkte
Materialien
white (polyamide, glass fibre and HPPE fibre steel)
white gloves
Größe
M
Sterilität
non-sterile
Leistungsmerkmale
latex free
powder-free
reusable
standard type EN388 (cat. II)
Verpackung
pack of 1 pair
Hersteller/Markenname
KCL 927
L × Dicke
250 mm × 1.50 mm
Allgemeine Beschreibung
The combination of polyamide, glass fiber and high tensile PE fiber guarantees a high cut resistance without lowering the gloves dexterity.
Nevocut is ambidextrous and can be washed several times without loosing its excellent fit.
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z677159-1PR | 4061833069004 |