Merck
Z674346
Duran® GLS 80 caps and accessories
Insert for GL 18 screw cap, I.D. 8.0 mm
Synonym(e):
1160171
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Hersteller/Markenname
DURAN® 1160171
Allgemeine Beschreibung
Flexible modular liquid or gas transfer 80 connection cap system. Five different tubing diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be connected. Sterile pressure equalisation is possible by using the syringe membrane filter. Unused ports can be closed with a closed top GL 18 PBT cap.
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)