Z673013
MSA disposable respirator for non-toxic dust
pkg of 50 ea
Alle Fotos(1)
About This Item
UNSPSC-Code:
46182002
NACRES:
NB.35
Empfohlene Produkte
Verpackung
pkg of 50 ea
Hersteller/Markenname
Conney Safety SAS2985
Verfügbarkeit
not available in EU
Allgemeine Beschreibung
Provides relief and protection from nuisance dusts such as animal dander, pollen, and other nuisance-level dusts at exposures below the OSHA Permissible Exposure Limit (PEL). Mask features an adjustable soft metal nosepiece and elastic headband. Not NIOSH approved.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)