Z616907
Vacuum accessories for Buchi® vacuum pumps
Needle valve gauge
Alle Fotos(1)
About This Item
UNSPSC-Code:
41104804
NACRES:
NB.43
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
Büchi 047291
Allgemeine Beschreibung
The simplest way to maintain vacuum control is through this cost-effective needle valve. Air is drawn in through a fine regulation valve and the resulting vacuum is set. A manometer displays the pressure. A Woulff bottle is recommended with this valve so that it can be a buffer vessel and condensate trap.
Rechtliche Hinweise
Büchi is a registered trademark of Büchi Labortechnik AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z616907-1EA | 4061832773742 |