Z569704
Wheaton cap for 1 L HDPE bottle
pkg of 800 ea
Synonym(e):
W083312B
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Empfohlene Produkte
Verpackung
pkg of 800 ea
Hersteller/Markenname
WHEATON®
Verwandte Kategorien
Leistungsmerkmale und Vorteile
- Double bagged twist-tied for reduced particulate
- Closures made of radiation grade Polypropylene
- All colorants meet CONEG regulations
- Methods of sterilization: gas, irradiation and disinfectant
- Closures packed separately and included with bottles
- Custom configurations available upon request
- Lot controlled
Rechtliche Hinweise
WHEATON is a registered trademark of Wheaton Industries, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z569704-800EA | 4061832831305 |