Z564869
Secador® Techni-Dome® 360 glove chamber
unobstructed viewing from any angle
Synonym(e):
humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
Alle Fotos(1)
About This Item
UNSPSC-Code:
41103406
NACRES:
NB.35
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art F50030-0000
Öffnung AD
6 in.
Volumen
65 L
65000 mL
Gewicht
5 kg
Allgemeine Beschreibung
The Techni-Dome® Glove Chamber offers generous interior workspace with a small footprint that preserves benchtop space. Each Techni-Dome® 360 Glove Chamber is molded of transparent polycarbonate for clear viewing from any angle. Polycarbonate construction affords resistance to a wide range of chemicals, high-impact strength and scratch-resistance and the smooth, hemispherical interior cleans easily.
Molded-in handles and low weight offer portability, making the unit suitable for field use. When no longer needed, the Glove Chamber can be quickly opened up, the two halves nested one inside the other, and the entire unit can be stowed away, regaining benchtop space.
The unit′s two-piece construction allows for quick processing of samples. To use the Glove Chamber, simply open the top half, place materials on the included platform, and close the unit.
The Glove Chamber is now ready for use. Molded-in ports on top and bottom and one included stopcock with plug can be used to purge the glove chamber with nitrogen or other gases to create a controlled atmosphere.
port clamps and gloves are not included
Molded-in handles and low weight offer portability, making the unit suitable for field use. When no longer needed, the Glove Chamber can be quickly opened up, the two halves nested one inside the other, and the entire unit can be stowed away, regaining benchtop space.
The unit′s two-piece construction allows for quick processing of samples. To use the Glove Chamber, simply open the top half, place materials on the included platform, and close the unit.
The Glove Chamber is now ready for use. Molded-in ports on top and bottom and one included stopcock with plug can be used to purge the glove chamber with nitrogen or other gases to create a controlled atmosphere.
port clamps and gloves are not included
Verlinkung
Full view of the Technidome.
Rechtliche Hinweise
Secador is a registered trademark of SP Scienceware, a division of SP
Techni-Dome is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
Z564869-1EA | 4061832265834 |