Z563978
Büchi® heating bath for R-210/215 evaporators
AC/DC input 220 - 240 V AC
Alle Fotos(1)
About This Item
UNSPSC-Code:
41100000
Empfohlene Produkte
Beschreibung
Model B-491
Hersteller/Markenname
Büchi 048200
Parameter
20-180 °C temp. limit
Kapazität
≤4 L
Allgemeine Beschreibung
The heating bath, for water or oil covers a wide temperature range. A continuous display shows the actual temperature and the temperature setpoint. It has a cordless power supply so it is not necessary to unplug connections when filling or emptying the bath.
Rechtliche Hinweise
Büchi is a registered trademark of Büchi Labortechnik AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)