Merck
Z549908
Sigma-Aldrich
Aldrich® jacketed three-neck flask
full jacket, capacity 1,000 mL, center joint: ST/NS 29/42
Synonym(e):
boiling flask, flask round, flask round bottom
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
borosilicate glass
round bottom
three-neck
three-neck flask
Verbindung
center joint: ST/NS29/42
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard Taper Joint
Verpackung
pack of 1
Hersteller/Markenname
Sigma-Aldrich
Kapazität
1,000 mL
Allgemeine Beschreibung
Unique design which incorporates a flush-closing PTFE drain valve eliminating unwanted dead space.
Features:
  • Eliminates dead space for complete stirring of flask contents
  • Allows for complete draining without breaking down flask set-up
  • Liquid cooled or heated with liquid or steam
  • 24/40 side joints
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z549908-1EA4061832133058