Z549819
Aldrich® jacketed three-neck flask
half jacket, capacity 1,000 mL, center joint: ST/NS 29/42
Synonym(e):
boiling flask, flask round, flask round bottom
About This Item
Empfohlene Produkte
Materialien
borosilicate glass
round bottom
three-neck
three-neck flask
Verbindung
center joint: ST/NS29/42
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard Taper Joint
Verpackung
pack of 1
Hersteller/Markenname
Sigma-Aldrich
Kapazität
1,000 mL
Verwandte Kategorien
Allgemeine Beschreibung
Features:
- Eliminates dead space for complete stirring of flask contents
- Allows for complete draining without breaking down flask set-up
- Liquid cooled or heated with liquid or steam
- 24/40 side joints
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z549819-1EA | 4061832118161 |