Merck
Z513369
Sigma-Aldrich
Mikrospatel
flat rounded/tapered ends, L 8 in. (203 mm)
About This Item
UNSPSC-Code:
41122403
NACRES:
NB.35
Materialien
nickel-stainless steel
Verpackung
pkg of 12 ea
Länge
8 in. (203 mm)
Allgemeine Beschreibung
Nickel stainless spatula with one rounded and one tapered end. Rounded end approximately 5/16 in. (8 mm) wide x 2 in. (51 mm) long. Tapered end approximately 1/4 in. (6 mm) wide x 1 3/4 in. (44 mm) long.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z513369-1PAK4061833580868