Z413577
Aldrich® three-way adapter with thermometer joint
joint: ST/NS 29/32
Synonym(e):
adapter laboratory, glass adapter, glass adapters
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
Empfohlene Produkte
Verbindung
thermometer joint ST/NS10/30
joint: ST/NS29/32
Verwandte Kategorien
Allgemeine Beschreibung
Side-arm is at 75° angle from bottom joint. Distance from joint to side-arm is 75mm.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z413577-1EA | 4061832115290 |