Merck
Z300993
Quickfit® Reduktionsadapter
top female joint: ST/NS 24/29, bottom male joint: ST/NS 45/40
Synonym(e):
DA37
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
Verbindung
female joint: ST/NS top24/29
male joint: ST/NS bottom45/40
Hersteller/Markenname
Quickfit DA 37
Verfügbarkeit
not available in USA and Canada
Allgemeine Beschreibung
These adapters connect items with a smaller cone to a piece of glassware with a larger size socket.
Rechtliche Hinweise
Quickfit is a registered trademark of SciLabware Limited
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)