Merck
Z300853
Quickfit® Reduktionsadapter
top female joint: ST/NS 10/19, bottom male joint: ST/NS 14/23
Synonym(e):
DA01
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
Verbindung
female joint: ST/NS top10/19
male joint: ST/NS bottom14/23
Hersteller/Markenname
Quickfit DA 01
Verfügbarkeit
not available in USA and Canada
Allgemeine Beschreibung
These adapters connect items with a smaller cone to a piece of glassware with a larger size socket.
Rechtliche Hinweise
Quickfit is a registered trademark of SciLabware Limited
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)