Z279994
Norprene® A-60-G Laborschlauch
I.D. × O.D. 3/8 in. × 9/16 in.
Synonym(e):
Saint Gobain AFL00028, Tygon A-60-G
Alle Fotos(1)
About This Item
UNSPSC-Code:
41105108
NACRES:
NB.51
Empfohlene Produkte
Verpackung
pkg of 50 feet
Hersteller/Markenname
Tygon
ID × AD
3/8 in. × 9/16 in.
Wandabmessungen
3/32 in.
Verwandte Kategorien
Allgemeine Beschreibung
This chemical pump tubing is a thermoplastic elastomer tubing with chemical compatibilities approaching fluoroelastomers and silicone tubing. Black, rubber-like tubing is heat sealable. Max. temp.: 275°F (135°C).
- Chemical resistance to oxidizing agents, acids, and alkalies
- Resistance to flex-fatigue, will not weaken or crack
Rechtliche Hinweise
Norprene is a registered trademark of Saint-Gobain Abrasives, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z279994-1PAK | 4061833578629 |