Z257575
PROHEAT® heat gun
PH-2100, AC/DC input 220 V AC, 3 - 6 A, Schuko plug
Alle Fotos(1)
About This Item
UNSPSC-Code:
41102404
NACRES:
NB.76
Empfohlene Produkte
Materialien
plastic
Leistungsmerkmale
CE compliant
Parameter
221-449 °C
453 L/min flow rate
Ausgabe
750-1300 W
Allgemeine Beschreibung
Compact, lightweight heat guns with dual temperature settings of 430°F and 840°F (221°C and 449°C). Excellent source of flameless heat with air flow at 15 cfm. The PROHEAT heat guns meet most general laboratory applications.
Rechtliche Hinweise
PROHEAT is a registered trademark of Master Appliance Corp.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z257575-1EA | 4061833293690 |