Merck
Z190276
Sigma-Aldrich
Arrow electric lab stirrer
speed 0-1750 rpm, direct, output0.066 hp
Synonym(e):
lab mixer, lab mixers, mixer stirrer
About This Item
UNSPSC-Code:
41103801
NACRES:
NB.72
Aufnahme AC/DC
115 V
115 V
Verpackung
pack of 1 ea
Hersteller/Markenname
Arrow Engineering 1750
Verfügbarkeit
not available in EU
Parameter
0-1750 rpm speed
Ausgabe
0.066 hp
Allgemeine Beschreibung
Totally enclosed, non-ventilated motor seals out corrosive fumes and dust. With on/off switch, circuit-breaker, and variable speed control. Includes a 9/16 × 10 in. support rod and 3/8 diam. × 12 in. L SS shaft and propeller.
Qualität
Note: Arrow lab stirrers conform to CSA22.2 but are not certified
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)