Merck
WHA2017013
Whatman® Antibiotic Assay Discs
diam. 13 mm
Synonym(e):
Whatman paper, Z741337
About This Item
UNSPSC-Code:
41111700
NACRES:
NB.79
Verpackung
pkg of 1000 ea
Hersteller/Markenname
Whatman 2017-013
Whatman Article No. 28418662 (US reference)
Durchmesser
13 mm
Allgemeine Beschreibung
For determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method.
Leistungsmerkmale und Vorteile
  • The antibiogram allows rational and selective chemotherapy.
  • Test discs can be coated with chemotherapeutic agents, placed on the innoculated nutrient agar and incubated.
  • The size of the inhibition zone is a measure for the effectiveness of the substances.
Sonstige Hinweise
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)