WHA10477601
Whatman® Vacuum filtration apparatus accessories
WT 100 Witt′s flask, with tubing nozzle, capacity
Synonym(e):
Z751960
Alle Fotos(1)
About This Item
UNSPSC-Code:
41104900
NACRES:
NB.24
Empfohlene Produkte
Verpackung
pk of 1
Hersteller/Markenname
Whatman 10477601
Whatman Article No. 28415990 (US reference)
Kapazität
Verwandte Kategorien
Allgemeine Beschreibung
Borosilicate glass bottle for filtrate collection in an inserted container.
Leistungsmerkmale und Vorteile
- For filtrate collection in an inserted container.
- The bottle is made of borosilicate glass.
- Has a replaceable round lid and side-mounted tubing nozzle for vacuum tubing 8 mm (inside diameter).
Sonstige Hinweise
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)