Merck
WHA10451610
Whatman® pressure filtration devices
MD 142/5/3 Pressure filtration apparatus, stainless steel, 2200 mL infusion vessel, 142 mm
Synonym(e):
Z751278
About This Item
UNSPSC-Code:
41104900
NACRES:
NB.24
Materialien
stainless steel
stainless steel housing
Leistungsmerkmale
2200 mL infusion vessel, 142 mm
Verpackung
pk of 1
Hersteller/Markenname
Whatman 10451610
Whatman Article No. 28415692 (US reference)
Allgemeine Beschreibung
Pressure filtration devices with a sample loading cylinder are suitable for batch filtration of samples from 20 ml. Devices without infusion cylinders are connected inline and are suitable for larger volumes of several liters.
Filtration of liquids and gases is possible, including sterile filtration* of serums or clear filtration of media that are difficult to filter, especially those that are highly viscous.
Leistungsmerkmale und Vorteile
  • Membranes, paper or glass fiber filter discs can be used.
  • Cleaning and changing of filters is completed in a few steps.
Sonstige Hinweise
Field of Use: For internal research use only. Products are not intended for diagnostic use or resale.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)