Merck
TEM-CFGA100CU
Sigma-Aldrich
Carbon Film Supported Copper Single Hole Grid
hole size 100 μm, box of 25
Synonym(e):
CARBON FILM ONLY SINGLE HOLE 100 MICRON COPPER GRIDS 25/BX
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Verpackung
box of 25
Lochgröße
100 μm
Allgemeine Beschreibung
A 5-6 nm thin film of pure carbon supported single hole grid. Copper grid, 100 micron hole. Carbon film show better electrical and heat conduction in the electron beam.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)