SYNV130501
Synthware™ conical bottom reaction vial with holed compression cap
0.1 mL, joint: ST/NS 5/5
Synonym(e):
Synthware V130501, reaction tube
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Empfohlene Produkte
Verbindung
joint: ST/NS5/5
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware V130501
Kapazität
0.1 mL
Verwandte Kategorien
Allgemeine Beschreibung
Supplied with one holed compression cap, one Viton® O-ring and one PTFE -faced silicone rubber septum. For replacement caps, see RC112; For Viton® O-rings, see R014; for septa, see R1420.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
SYNV130501-1EA | 4061832198804 |