Merck
SYNP160006
Synthware cylindrical pressure vessel, heavy wall
120 mL, O.D. × L 46 mm × 169 mm, #15 bushing
Synonym(e):
Synthware P160006, pressure tube
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.51
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware P160006
AD × L
46 mm × 169 mm
Kapazität
120 mL
Fitting
#15 bushing
Allgemeine Beschreibung
Heavy wall cylindrical vessels have an internal thread for use with a 15 mm PTFE bushing as a pressure seal. This unique design allows hand tightening and loosening for easy access into the vessel. The O-ring is positioned on the bushing above the thread for improved sealing. The vessels are useful for pressure reactions and applications such as polymerization and analysis of halogens by thermal decomposition in the presence of lime. Complete apparatus is supplied with vessel, PTFE bushing and Viton® O-ring. *Important Warnings for Pressurized Glassware: Due to varying conditions, we cannot guarantee glass vessels from breakage under pressure. Always work behind a safety shield.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)