Merck
SYNM490064
Synthware vacuum manifold with PTFE stopcocks
number of ports, 6, Hose Connection: left-right, stopcock size 4 mm
Synonym(e):
Synthware M490064, vaccum unit
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
Leistungsmerkmale
number of ports 6
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware M490064
Absperrhahngröße
4 mm
Allgemeine Beschreibung
General purpose manifold with 2 mm or 4 mm PTFE stopcocks. Main tube is 22 mm O.D. with serrated hose connection(10 mm O.D.) at both ends. Delivery ports have 10 mm serrated hose connections. Distance between stopcocks is75 mm.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)