SYNM290005
Synthware™ all-glass vacuum/inert gas manifold with solid high vacuum stopcocks
number of ports, 5, Hose Connections: Front-left-right, Rear-right
Synonym(e):
Synthware M290005, vacuum manifold, vacuum unit
Alle Fotos(1)
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
Empfohlene Produkte
Leistungsmerkmale
number of ports 5
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware M290005
Verwandte Kategorien
Allgemeine Beschreibung
With 4 mm double oblique, high vacuum solid glass stopcocks precision ground with corresponding numbers on barrel and plug. Solid glass stopcocks have constant pressure retention devices. Beams are 22 mm O.D. with front-left-right and rear-right 10 mm hose connections. Distance between stopcocks is75 mm center to center.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)