SYNF963014
Synthware™Buchner funnel with perforated plate
30 mL, joint: ST/NS 14/20, plate O.D. 32 mm
Synonym(e):
Synthware F963014, filter funnel
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.43
Empfohlene Produkte
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F963014
Kapazität
30 mL
30 mL
Platte AD
32 mm
Verwandte Kategorien
Allgemeine Beschreibung
Synthware™ Büchner funnel with an inner joint and a perforated glass plate, sealed with a perforated glass disc to support filter media. It features a standard taper inner joint (14/20) and a hose connector for the vacuum source.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
SYNF963014-1EA | 4061832196701 |